Merck
  • Over-expression of miR-411-5p and miR-434-3p promotes the osteoblast differentiation by targeting GATA4.
Over-expression of miR-411-5p and miR-434-3p promotes the osteoblast differentiation by targeting GATA4.
Molecular and cellular endocrinology (2020-02-18)
Xuren Gao, Jian Ge, Weiyi Li, Wang-Chen Zhou, Lei Xu, De-Qin Geng
ABSTRACT
To investigate the role of miR-411-5p and miR-434-3p in osteoblast differentiation in particulate-induced osteolysis. A mouse model of osteolysis and an in vitro osteolysis model were constructed. The expressions of molecules were detected using qRT-PCR and western blot. Alkaline phosphatase (ALP) activity was measured using the ALP Assay Kit, and the bone mineralization was measured using alizarin red staining. The expression of miR-411-5p and miR-434-3p was decreased in osteolysis mice and UHMWPE-induced mMSCs, while GATA4 protein expression was increased. Over-expression of miR-411-5p and miR-434-3p up-regulated the expressions of osteoblast gene markers, enhanced the ALP activity, promoted the bone mineralization of mesenchymal stem cells. In addition, miR-411-5p and miR-434-3p could target GATA4, and miR-411-5p/434-3p affected the expressions of osteoblast gene markers through GATA4 in vitro and in vivo. Overexpression of miR-411-5p and miR-434-3p promoted the osteoblast differentiation by inhibiting GATA4 expression.
Sigma-Aldrich
MISSION® esiRNA, targeting human GATA4
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)