Merck
  • Focal adhesion-localization of START-GAP1/DLC1 is essential for cell motility and morphology.
Focal adhesion-localization of START-GAP1/DLC1 is essential for cell motility and morphology.
Genes to cells : devoted to molecular & cellular mechanisms (2009-01-28)
Katsuhisa Kawai, Yui Iwamae, Masaki Yamaga, Minoru Kiyota, Hiroko Ishii, Hajime Hirata, Yoshimi Homma, Hitoshi Yagisawa
ABSTRACT
There is a class of GTPase activating proteins for the Rho family GTPases (RhoGAPs) that contain the steroidogenic acute regulatory protein (STAR)-related lipid transfer (START) domain. In mammals three genes encode such proteins and they are designated START-GAP1-3 or deleted in liver cancer 1-3 (DLC1-3). In this study, we examined the intracellular localization and roles of START-GAP1/DLC1 in cell motility. Immunofluorescence microscopic analysis of NRK cells and HeLa cells revealed that START-GAP1 was localized in focal adhesions. Amino acid residues 265-459 of START-GAP1 were found to be necessary for focal adhesion targeting and we name the region "the focal adhesion-targeting (FAT) domain." It was previously known that ectopic expression of START-GAP1 induced cell rounding. We demonstrated that the FAT domain of START-GAP1 was partially required for this morphological change. Furthermore, expression of this domain in HeLa cells resulted in dissociation of endogenous START-GAP1 from focal adhesions as a dominant negative modulator, reducing cell migration and spreading. Taken together, START-GAP1 is targeted to focal adhesions via the FAT domain and regulates actin rearrangement through down-regulation of active RhoA and Cdc42. Its absence from focal adhesions could, therefore, cause abnormal cell motility and spreading.