Merck
  • Separation of carbamazepine and five metabolites, and analysis in human plasma by micellar electrokinetic capillary chromatography.
Separation of carbamazepine and five metabolites, and analysis in human plasma by micellar electrokinetic capillary chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2002-05-16)
Maria Augusta Raggi, Vincenzo Pucci, Alessandra Maurizio, Jan Muzikar, Ernst Kenndler
ABSTRACT
A rapid and feasible method was developed for the analysis of carbamazepine and its five metabolites (10,11-dihydro-10,11-epoxycarbamazepine, 10,11-dihydro-10,11-dihydroxycarbamazepine, 10,11-dihydro-10-hydroxycarbamazepine, 2-hydroxycarbamazepine and 3-hydroxycarbamazepine) in human plasma. Separation of the analytes is based on micellar electrokinetic chromatography, in untreated fused-silica capillary (48.5/40.0 cm length, 50 microm I.D.) with phosphate buffer (30 mM, pH 8.00) as background electrolyte, containing 50 mM sodium dodecylsulfate, and methanol (15%, v/v) as organic modifier. Clean up of human plasma samples was carried out by means of a solid-phase extraction procedure, which gave a high extraction yield for all six carbamazepines (>88%). The overall precision of the method gives a mean RSD of about 1.8%. The limit of quantitation for all analytes is < or = 0.30 microg ml(-1), the limit of detection < or = 0.12 microg ml(-1).