Merck
  • Uncoupling endosomal CLC chloride/proton exchange causes severe neurodegeneration.
Uncoupling endosomal CLC chloride/proton exchange causes severe neurodegeneration.
The EMBO journal (2020-03-03)
Stefanie Weinert, Niclas Gimber, Dorothea Deuschel, Till Stuhlmann, Dmytro Puchkov, Zohreh Farsi, Carmen F Ludwig, Gaia Novarino, Karen I López-Cayuqueo, Rosa Planells-Cases, Thomas J Jentsch
ABSTRACT
CLC chloride/proton exchangers may support acidification of endolysosomes and raise their luminal Cl- concentration. Disruption of endosomal ClC-3 causes severe neurodegeneration. To assess the importance of ClC-3 Cl- /H+ exchange, we now generate Clcn3unc/unc mice in which ClC-3 is converted into a Cl- channel. Unlike Clcn3-/- mice, Clcn3unc/unc mice appear normal owing to compensation by ClC-4 with which ClC-3 forms heteromers. ClC-4 protein levels are strongly reduced in Clcn3-/- , but not in Clcn3unc/unc mice because ClC-3unc binds and stabilizes ClC-4 like wild-type ClC-3. Although mice lacking ClC-4 appear healthy, its absence in Clcn3unc/unc /Clcn4-/- mice entails even stronger neurodegeneration than observed in Clcn3-/- mice. A fraction of ClC-3 is found on synaptic vesicles, but miniature postsynaptic currents and synaptic vesicle acidification are not affected in Clcn3unc/unc or Clcn3-/- mice before neurodegeneration sets in. Both, Cl- /H+ -exchange activity and the stabilizing effect on ClC-4, are central to the biological function of ClC-3.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-GABAA Receptor α1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PSD-95 (Ab-2) Mouse mAb (7E3-1B8), liquid, clone 7E3-1B8, Calbiochem®