Merck
  • An improved method for the synthesis of protected glycosyl fluorides from thioglycosides using N,N-diethylaminosulfur trifluoride (DAST).
An improved method for the synthesis of protected glycosyl fluorides from thioglycosides using N,N-diethylaminosulfur trifluoride (DAST).
Carbohydrate research (2012-08-29)
Katsuhiko Suzuki, Yukishige Ito, Osamu Kanie
ABSTRACT
The direct conversion of thioglycosides to glycosyl fluorides frequently used in oligosaccharide synthesis was examined using N,N-diethylaminosulfur trifluoride (DAST). Although the reaction proceeded without N-bromosuccinimide (NBS), in some cases it was found that the electrophilicity of the Vilsmeier-type electrophilic sulfinium cation species was not sufficient for the activation of certain less-reactive thioglycosides. Here, we report the results of fluorination reactions of a series of monosaccharides using DAST in the absence of NBS, and also discuss the acceleration of the reaction in the presence of dimethyl(methylthio)sulfonium trifluoromethanesulfonate (DMTST) resulting in excellent product yields.
Sigma-Aldrich
(Diethylamino)sulfur trifluoride, 95%