Merck
  • A remarkably efficient fluoroalkylation of cyclic sulfates and sulfamidates with PhSO2CF2H: facile entry into beta-difluoromethylated or beta-difluoromethylenated alcohols and amines.
A remarkably efficient fluoroalkylation of cyclic sulfates and sulfamidates with PhSO2CF2H: facile entry into beta-difluoromethylated or beta-difluoromethylenated alcohols and amines.
Angewandte Chemie (International ed. in English) (2006-12-14)
Chuanfa Ni, Jun Liu, Laijun Zhang, Jinbo Hu
Sigma-Aldrich
Difluoromethyl phenyl sulfone, ≥97%