Merck
  • Lipid-Sorting Specificity Encoded in K-Ras Membrane Anchor Regulates Signal Output.
Lipid-Sorting Specificity Encoded in K-Ras Membrane Anchor Regulates Signal Output.
Cell (2017-01-04)
Yong Zhou, Priyanka Prakash, Hong Liang, Kwang-Jin Cho, Alemayehu A Gorfe, John F Hancock
ABSTRACT
K-Ras is targeted to the plasma membrane by a C-terminal membrane anchor that comprises a farnesyl-cysteine-methyl-ester and a polybasic domain. We used quantitative spatial imaging and atomistic molecular dynamics simulations to examine molecular details of K-Ras plasma membrane binding. We found that the K-Ras anchor binds selected plasma membrane anionic lipids with defined head groups and lipid side chains. The precise amino acid sequence and prenyl group define a combinatorial code for lipid binding that extends beyond simple electrostatics; within this code lysine and arginine residues are non-equivalent and prenyl chain length modifies nascent polybasic domain lipid preferences. The code is realized by distinct dynamic tertiary structures of the anchor on the plasma membrane that govern amino acid side-chain-lipid interactions. An important consequence of this specificity is the ability of such anchors when aggregated to sort subsets of phospholipids into nanoclusters with defined lipid compositions that determine K-Ras signaling output.
Product Description
Avanti
18:0-18:1 PS, 1-stearoyl-2-oleoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Pricing and availability is not currently available.