Merck
  • CUB Domain-Containing Protein-1 Promotes Proliferation, Migration and Invasion in Cervical Cancer Cells.
CUB Domain-Containing Protein-1 Promotes Proliferation, Migration and Invasion in Cervical Cancer Cells.
Cancer management and research (2020-06-18)
Lijun Huang, Yihong Chen, Shuyu Lai, Hongmei Guan, Xiaoling Hu, Jie Liu, Hanrong Zhang, Zhenfei Zhang, Jueyu Zhou
ABSTRACT
Emerging evidence have revealed significant contributions of CUB domain-containing protein-1 (CDCP1) in tumorigenesis, including colon, renal, ovarian, pancreatic, prostate and breast cancers. However, the roles of CDCP1 in cervical cancer (CC) still remain elusive. Quantitative reverse transcription polymerase chain reaction, immunohistochemistry and Western blotting were used to confirm the expression of CDCP1 in CC tissues compared with matched non-tumor tissues. In vitro, gain-of-function and loss-of-function studies were used to investigate the biological function and underlying mechanism of CDCP1 in cervical carcinogenesis. Furthermore, tumor growth was evaluated using a xenogenous subcutaneously implant model of CC cells in vivo. Here, we confirmed that CDCP1 was significantly increased in human CC both in mRNA and in protein levels compared to normal cervical tissues. Furthermore, we demonstrated that increased CDCP1 expression promotes proliferation, migration, invasion and mediates the epithelial-to-mesenchymal transition phenotype in HeLa and C33A cells. Also, CDCP1 knockdown reverses all the effects of enhanced CDCP1 on cell behavior in SiHa and Caski cells. Importantly, the suppressive expression of CDCP1 repressed tumor growth in a mouse xenograft model of CC. In summary, our current study results provide novel insights into the role of CDCP1 in CC progression. Potentially, CDCP1 might serve as a diagnostic biomarker and a novel therapeutic target for CC.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1
Sigma-Aldrich
MISSION® esiRNA, targeting human CDCP1
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile