Merck
  • Prophylactic low-molecular-weight heparin administration protected against severe acute pancreatitis partially by VEGF/Flt-1 signaling in a rat model.
Prophylactic low-molecular-weight heparin administration protected against severe acute pancreatitis partially by VEGF/Flt-1 signaling in a rat model.
Human & experimental toxicology (2020-05-01)
S Li, S Zhang, R Li, S Chen, S Chang, X Chen, Y Li, X Su, T Wu, M Xu
ABSTRACT
The present study was aimed to explore the effects and the underlying mechanism of prophylactic low-molecular-weight heparin (LMWH) treatment on taurocholate-induced severe acute pancreatitis (SAP) in a rat model. Rat SAP model was induced by injection of 4% sodium taurocholate into the pancreatic duct. LMWH was applied half an hour before the induction of pancreatitis at the dose of 200 IU/kg subcutaneous injection. The rats were euthanized at 1 h, 6 h, and 12 h after taurocholate-induced SAP. The inflammatory and oxidative response markers were assessed. And the vascular endothelial growth factor (VEGF) and Fms-related tyrosine kinase 1 (Flt-1) expression were evaluated by immunohistochemistry (IHC) and western blot methods. The expression of inflammatory and oxidative response markers increased after induction of SAP. IHC and western blot results showed the VEGF and Flt-1 expression were increased in SAP group. Prophylactic LMWH administration reduced the inflammatory and oxidative response markers expression and decreased the expression of VEGF and Flt-1. This study suggested that prophylactic LMWH treatment mitigated the severity of pancreatitis in rat SAP model by anti-inflammation and oxidative response. The underlying mechanism may result from downregulating VEGF/Flt-1 signaling of LMWH in SAP rat model.
Product Description
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard