Merck
  • Deletion of atypical chemokine receptor 3 (ACKR3) increases immune cells at the fetal-maternal interface.
Deletion of atypical chemokine receptor 3 (ACKR3) increases immune cells at the fetal-maternal interface.
Placenta (2020-05-27)
Kelsey E Quinn, Brooke C Matson, Kathleen M Caron
ABSTRACT
Establishment of immune cell populations and adaptations in immune cells are critical aspects during pregnancy that lead to protection of the semi-allogenic fetus. Appropriate immune cell activation and trophoblast migration are regulated in part by chemokines, the availability of which can be fine-tuned by decoy receptors. Atypical chemokine receptor 3 (ACKR3), previously named C-X-C chemokine receptor 7 (CXCR7), is a chemokine decoy receptor expressed in placenta, but little is known about how this receptor affects placental development. In this study, we investigated the phenotypic characteristics of placentas from Ackr3-/- embryos to determine how Ackr3 contributes to early placentation. In placentas from Ackr3-/- embryos, we observed an increase in decidual compaction and in the size of the uterine natural killer cell population. Ackr3 knockdown in trophoblast cells led to a decrease in trophoblast migration. These findings suggest that this decoy receptor may therefore be an important factor in normal placentation.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cxcr7
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)