Merck
  • Extraction of Microfibrillar Cellulose From Waste Paper by NaOH/Urethane Aqueous System and Its Utility in Removal of Lead from Contaminated Water.
Extraction of Microfibrillar Cellulose From Waste Paper by NaOH/Urethane Aqueous System and Its Utility in Removal of Lead from Contaminated Water.
Materials (Basel, Switzerland) (2020-07-08)
Vadahanambi Sridhar, Hyun Park
ABSTRACT
Though recycling of waste paper is widely practiced but usually it is downgraded to lower valued recycled waste paper. Based on this concern, we report the development of novel NaOH/urethane aqueous system for extraction of microfibrillated cellulose from waste paper. The purity of so obtained microfibrillated cellulose (MFC) was evaluated by morphological tests using scanning electron microscopy (SEM) and transmission electron microscopy (TEM) and by evaluation of physicochemical properties using Fourier-transform infrared (FTIR) spectroscopy, X-ray diffraction (XRD), differential scanning calorimetry (DSC), and thermogravimetric analysis (TGA). Morphologies of MFC studied by SEM and TEM showed that the size of purified cellulose fibrils reduced when compared to that of waste paper but fibrils are cleaner and smoother due to the removal of talc and lignin. XRD analysis revealed that MFC exhibits good crystallinity. The utility of sulfonated and pristine microfibrillar cellulose in removal of lead from contaminated water is also reported. Our results show that renewable, sustainable, cheap, and waste biomass like waste paper can be used for producing valuable second-generation high-value products.
Sigma-Aldrich
D-(+)-Talose, ≥99.0% (HPLC)
Sigma-Aldrich
p-Toluenesulfonyl chloride, reagent grade, ≥98%
Sigma-Aldrich
N,N-Dimethylacetamide-d9, 99 atom % D