Merck
  • A New Conducting Copolymer Bearing Electro-Active Nitroxide Groups as Organic Electrode Materials for Batteries.
A New Conducting Copolymer Bearing Electro-Active Nitroxide Groups as Organic Electrode Materials for Batteries.
ChemSusChem (2020-04-22)
L Assumma, Y Kervella, J-M Mouesca, M Mendez, V Maurel, L Dubois, T Gutel, S Sadki
ABSTRACT
To reduce the amount of conducting additives generally required for polynitroxide-based electrodes, a stable radical (TEMPO) is combined with a conductive copolymer backbone consisting of 2,7-bisthiophene carbazole (2,7-BTC), which is characterized by a high intrinsic electronic conductivity. This work deals with the synthesis of this new polymer functionalized by a redox nitroxide. Fine structural characterization using electron paramagnetic resonance (EPR) techniques established that: 1) the nitroxide radicals are properly attached to the radical chain (continuous wave EPR) and 2) the polymer chain has very rigid conformations leading to a set of well-defined distances between first neighboring pairs of nitroxides (pulsed EPR). The redox group combined with the electroactive polymer showed not only a very high electrochemical reversibility but also a perfect match of redox potentials between the de-/doping reaction of the bisthiophene carbazole backbone and the redox activity of the nitroxide radical. This new organic electrode shows a stable capacity (about 60 mAh g-1 ) and enables a strong reduction in the amount of carbon additive due to the conducting-polymer skeleton.
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidinol, 98%