Merck
  • Supplementation with omega-3 fatty acids potentiates oxidative stress in human airway epithelial cells exposed to ozone.
Supplementation with omega-3 fatty acids potentiates oxidative stress in human airway epithelial cells exposed to ozone.
Environmental research (2020-05-18)
Elizabeth M Corteselli, Avram Gold, Jason Surratt, Tianqu Cui, Philip Bromberg, Lisa Dailey, James M Samet
ABSTRACT
Dietary intake of the omega-3 family of polyunsaturated fatty acids (ω-3 FA) is associated with anti-inflammatory effects. However, unsaturated fatty acids are susceptible to oxidation, which produces pro-inflammatory mediators. Ozone (O3) is a tropospheric pollutant that reacts rapidly with unsaturated fatty acids to produce electrophilic and oxidative mediators of inflammation. Determine whether supplementation with ω-3 FA alters O3-induced oxidative stress in human airway epithelial cells (HAEC). 16-HBE cells expressing a genetically encoded sensor of the reduced to oxidized glutathione ratio (GSH/GSSG, EGSH) were supplemented with saturated, monounsaturated, or ω-3 FA prior to exposure to 0, 0.08, 0.1, or 0.3 ppm O3. Lipid peroxidation was measured in cellular lipid extracts and intact cells following O3 exposure. Relative to cells incubated with the saturated or monounsaturated fatty acids, cells supplemented with ω-3 FA containing 5 or 6 double bonds showed a marked increase in EGSH during exposure to O3 concentrations as low as 0.08 ppm. Consistent with this finding, the concentration of lipid hydroperoxides produced following O3 exposure was significantly elevated in ω-3 FA supplemented cells. Supplementation with polyunsaturated ω-3 FA potentiates oxidative responses, as indicated by EGSH, in HAEC exposed to environmentally relevant concentrations of O3. This effect is mediated by the increased formation of lipid hydroperoxides produced by the reaction of O3 with polyunsaturated fatty acids. Given the inflammatory activity of lipid hydroperoxides, these findings have implications for the potential role of ω-3 FA in increasing human susceptibility to the adverse health effects of O3 exposure.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)