Merck
  • CD147 is a Novel Interaction Partner of Integrin αMβ2 Mediating Leukocyte and Platelet Adhesion.
CD147 is a Novel Interaction Partner of Integrin αMβ2 Mediating Leukocyte and Platelet Adhesion.
Biomolecules (2020-04-08)
David Heinzmann, Moritz Noethel, Saskia V Ungern-Sternberg, Ioannis Mitroulis, Meinrad Gawaz, Triantafyllos Chavakis, Andreas E May, Peter Seizer
ABSTRACT
Surface receptor-mediated adhesion is a fundamental step in the recruitment of leukocytes and platelets, as well as platelet-leukocyte interactions. The surface receptor CD147 is crucially involved in host defense against self-derived and invading targets, as well as in thrombosis. In the current study, we describe the previously unknown interaction of CD147 with integrin αMβ2 (Mac-1) in this context. Using binding assays, we were able to show a stable interaction of CD147 with Mac-1 in vitro. Leukocytes from Mac-1-/- and CD147+/- mice showed a markedly reduced static adhesion to CD147- and Mac-1-coated surfaces, respectively, compared to wild-type mice. Similarly, we observed reduced rolling and adhesion of monocytes under flow conditions when cells were pre-treated with antibodies against Mac-1 or CD147. Additionally, as assessed by antibody inhibition experiments, CD147 mediated the dynamic adhesion of platelets to Mac-1-coated surfaces. The interaction of CD147 with Mac-1 is a previously undescribed mechanism facilitating the adhesion of leukocytes and platelets.