Merck
  • CENP-B creates alternative epigenetic chromatin states permissive for CENP-A or heterochromatin assembly.
CENP-B creates alternative epigenetic chromatin states permissive for CENP-A or heterochromatin assembly.
Journal of cell science (2020-07-15)
Koichiro Otake, Jun-Ichirou Ohzeki, Nobuaki Shono, Kazuto Kugou, Koei Okazaki, Takahiro Nagase, Hisashi Yamakawa, Natalay Kouprina, Vladimir Larionov, Hiroshi Kimura, William C Earnshaw, Hiroshi Masumoto
ABSTRACT
CENP-B binds to CENP-B boxes on centromeric satellite DNAs (known as alphoid DNA in humans). CENP-B maintains kinetochore function through interactions with CENP-A nucleosomes and CENP-C. CENP-B binding to transfected alphoid DNA can induce de novo CENP-A assembly, functional centromere and kinetochore formation, and subsequent human artificial chromosome (HAC) formation. Furthermore, CENP-B also facilitates H3K9 (histone H3 lysine 9) trimethylation on alphoid DNA, mediated by Suv39h1, at ectopic alphoid DNA integration sites. Excessive heterochromatin invasion into centromere chromatin suppresses CENP-A assembly. It is unclear how CENP-B controls such different chromatin states. Here, we show that the CENP-B acidic domain recruits histone chaperones and many chromatin modifiers, including the H3K36 methylase ASH1L, as well as the heterochromatin components Suv39h1 and HP1 (HP1α, β and γ, also known as CBX5, CBX1 and CBX3, respectively). ASH1L facilitates the formation of open chromatin competent for CENP-A assembly on alphoid DNA. These results indicate that CENP-B is a nexus for histone modifiers that alternatively promote or suppress CENP-A assembly by mutually exclusive mechanisms. Besides the DNA-binding domain, the CENP-B acidic domain also facilitates CENP-A assembly de novo on transfected alphoid DNA. CENP-B therefore balances CENP-A assembly and heterochromatin formation on satellite DNA.
Sigma-Aldrich
TN-16, ≥98% (HPLC)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)