Merck
  • Suberoyl bis-hydroxamic acid induces p53-dependent apoptosis of MCF-7 breast cancer cells.
Suberoyl bis-hydroxamic acid induces p53-dependent apoptosis of MCF-7 breast cancer cells.
Acta pharmacologica Sinica (2008-11-26)
Zhi-gang Zhuang, Fei Fei, Ying Chen, Wei Jin
ABSTRACT
To study the effects of suberoyl bis-hydroxamic acid (SBHA), an inhibitor of histone deacetylases, on the apoptosis of MCF-7 breast cancer cells. Apoptosis in MCF-7 cells induced by SBHA was demonstrated by flow cytometric analysis, morphological observation, and DNA ladder. Mitochondrial membrane potential (DeltaPsim) was measured using the fluorescent probe JC-1. The expressions of p53, p21, Bax, and PUMA were determined using RT-PCR or Western blotting analysis after the MCF-7 cells were treated with SBHA or p53 siRNA. SBHA induced apoptosis in MCF-7 cells. The expressions of p53, p21, Bax, and PUMA were induced, and DeltaPsim collapsed after treatment with SBHA. p53 siRNA abrogated the SBHA-induced apoptosis and the expressions of p53, p21, Bax, and PUMA. The activation of the p53 pathway is involved in SBHA-induced apoptosis in MCF-7 cells.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
SBHA, A cell-permeable bishydroxamic acid histone deacetylase (HDAC) inhibitor with anti-tumor properties.
Sigma-Aldrich
Suberohydroxamic acid, 95%