- Proteinase K from Tritirachium album Limber.
Proteinase K from Tritirachium album Limber.
European journal of biochemistry (1974-08-15)
W Ebeling, N Hennrich, M Klockow, H Metz, H D Orth, H Lang
PMID4373242
Sigma-Aldrich
Proteinase K from Tritirachium album, Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥30 units/mg protein, Molecular Biology
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL