- Synthesis of highly enantiomerically enriched amines by asymmetric transfer hydrogenation of N-(tert-butylsulfinyl) imines
Synthesis of highly enantiomerically enriched amines by asymmetric transfer hydrogenation of N-(tert-butylsulfinyl) imines
Organic Syntheses (2013)
Guijarro D, et al.