Merck
  • Carbonic anhydrase inhibitors: synthesis and inhibition of cytosolic/tumor-associated carbonic anhydrase isozymes I, II, and IX with sulfonamides incorporating thioureido-sulfanilyl scaffolds.
Carbonic anhydrase inhibitors: synthesis and inhibition of cytosolic/tumor-associated carbonic anhydrase isozymes I, II, and IX with sulfonamides incorporating thioureido-sulfanilyl scaffolds.
Bioorganic & medicinal chemistry letters (2005-04-20)
Luca Puccetti, Giuseppe Fasolis, Alessandro Cecchi, Jean-Yves Winum, Alessandro Gamberi, Jean-Louis Montero, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
The tumor-associated transmembrane carbonic anhydrase (CA, EC 4.2.1.1) isozyme IX (CA IX) is overexpressed in hypoxic tumors and appears to be involved in acidification of the tumor microenvironment, a process correlated with cancer progression and bad prognosis. The acidification may be reduced by inhibiting the enzyme with potent sulfonamide/sulfamate CA inhibitors. A series of such aromatic sulfonamides incorporating thioureido-sulfanilyl moieties has been prepared and investigated for its interaction with the catalytic domain of the human isozyme hCA IX. The key intermediates in the synthesis were obtained by reacting sulfanilamide, homosulfanilamide, or 4-aminoethylbenzenesulfonamide with 4-acetamido-benzenesulfonyl chloride followed by deacetylation and reaction with thiophosgene. The obtained isothiocyanato sulfonamides were reacted with aliphatic or aromatic primary amines or hydrazines, leading to the corresponding thioureas. Some of these compounds showed excellent inhibitory properties against isozymes I, II, and IX, with several inhibitors also presenting selectivity for the inhibition of CA IX over that of the ubiquitous isozyme CA II. Such sulfonamides may constitute interesting candidates for the development of novel antitumor therapies based on the inhibition of the CA isozymes overexpressed in hypoxic tumors. Due to the highest expression of CA IX in clear renal cell carcinoma and its chemo/radioresistance, our efforts are first of all directed to generate effective therapeutic strategies for the cure of this malignancy.
Product Description
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Sulfanilamide, ≥98%