Merck
  • The transgenic Arabidopsis plant system, pER8-GFP, as a powerful tool in searching for natural product estrogen-agonists/antagonists.
The transgenic Arabidopsis plant system, pER8-GFP, as a powerful tool in searching for natural product estrogen-agonists/antagonists.
Journal of natural products (2005-07-26)
Fang-Rong Chang, Ken-ichiro Hayashi, Nam-Hai Chua, Shuichi Kamio, Zih-You Huang, Hiroshi Nozaki, Yang-Chang Wu
ABSTRACT
The transgenic Arabidopsis plant system, pER8-GFP, may be used as a powerful tool in searching for natural estrogen-agonists/antagonists. Among selected plant extracts and natural products, the method was able to distinguish active extracts (e.g., Glycine max and Pueraria lobata) and pure compounds (e.g., 17beta-estradiol (1), genistein (10), and daidzein (11)) and also to distinguish effects of structural changes on activity. Thus, this rapid sensitive system was proven to be suitable for the discovery of natural products with estrogenic activity.
Product Description
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Cholecalciferol, analytical standard
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Supelco
Puerarin, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, analytical standard