Merck
  • Hydroxy-1,2,5-oxadiazolyl moiety as bioisoster of the carboxy function. Synthesis, ionization constants, and pharmacological characterization of gamma-aminobutyric acid (GABA) related compounds.
Hydroxy-1,2,5-oxadiazolyl moiety as bioisoster of the carboxy function. Synthesis, ionization constants, and pharmacological characterization of gamma-aminobutyric acid (GABA) related compounds.
Journal of medicinal chemistry (2006-07-11)
Marco L Lolli, Suzanne L Hansen, Barbara Rolando, Birgitte Nielsen, Petrine Wellendorph, Karsten Madsen, Orla Miller Larsen, Uffe Kristiansen, Roberta Fruttero, Alberto Gasco, Tommy N Johansen
ABSTRACT
Three 4-substituted 1,2,5-oxadiazol-3-ols containing aminoalkyl substituents (analogues and homologues of gamma-aminobutyric acid (GABA)) were synthesized to investigate the hydroxy-1,2,5-oxadiazolyl moiety as a bioisoster for a carboxyl group at GABA receptors. The pK(a) values of the target compounds were close to those of GABA. At GABA(A) receptors of cultured cerebral cortical neurons, weak agonist and partial agonist profiles were identified, demonstrating the 4-hydroxy-1,2,5-oxadiazol-3-yl unit to be a nonclassical carboxyl group bioisoster.
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%