Merck
  • Myelin and Lipid Composition of the Corpus Callosum in Mucopolysaccharidosis Type I Mice.
Myelin and Lipid Composition of the Corpus Callosum in Mucopolysaccharidosis Type I Mice.
Lipids (2020-06-17)
Steven Q Le, Igor Nestrasil, Shih-Hsin Kan, Martin Egeland, Jonathan D Cooper, David Elashoff, Rong Guo, Jakub Tolar, Jennifer K Yee, Patricia I Dickson, Steven Q Le, Igor Nestrasil, Shih-Hsin Kan, Martin Egeland, Jonathan D Cooper, David Elashoff, Rong Guo, Jakub Tolar, Jennifer K Yee, Patricia I Dickson
ABSTRACT
Mucopolysaccharidosis type I (MPS I) is a lysosomal disease with progressive central nervous system involvement. This study examined the lipid, cholesterol, and myelin basic protein composition of white matter in the corpus callosum of MPS I mice. We studied 50 week-old, male MPS I mice and littermate, heterozygote controls (n = 12 per group). Male MPS I mice showed lower phosphatidylcholine and ether-linked phosphatidylcholine quantities than controls (p < 0.05). Twenty-two phospholipid or ceramide species showed significant differences in percent of total. Regarding specific lipid species, MPS I mice exhibited lower quantities of sphingomyelin 18:1, phosphatidylserine 38:3, and hexosylceramide d18:1(22:1) mH2 O than controls. Principal components analyses of polar, ceramide, and hexosylceramide lipids, respectively, showed some separation of MPS I and control mice. We found no significant differences in myelin gene expression, myelin basic protein, or total cholesterol in the MPS I mice versus heterozygous controls. There was a trend toward lower proteolipid protein-1 levels in MPS I mice (p = 0.06). MPS I mice show subtle changes in white matter composition, with an unknown impact on pathogenesis in this model.
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MBP (myelin basic protein), bovine, purified
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid