Merck
  • Design, synthesis, and biological evaluation of Plasmodium falciparum lactate dehydrogenase inhibitors.
Design, synthesis, and biological evaluation of Plasmodium falciparum lactate dehydrogenase inhibitors.
Journal of medicinal chemistry (2007-07-20)
Seoung-ryoung Choi, Anupam Pradhan, Nicholas L Hammond, Amar G Chittiboyina, Babu L Tekwani, Mitchell A Avery
ABSTRACT
Plasmodium falciparum lactate dehydrogenase (pfLDH) is a key enzyme for energy generation of malarial parasites and is a potential antimalarial chemotherapeutic target. It is known that the oxamate moiety, a pyruvate analog, alone shows higher inhibition against pfLDH than human LDHs, suggesting that it can be used for the development of selective inhibitors. Oxamic acid derivatives were designed and synthesized. Derivatives 5 and 7 demonstrated activities against pfLDH with IC50 values of 3.13 and 1.75 muM, respectively, and have 59- and 7-fold selectivity over mammalian LDH, respectively. They also have micromolar range activities against Plasmodium falciparum malate dehydrogenase (pfMDH), which may fill the role of pfLDH when the activity of pfLDH is reduced. Thus, certain members of these oxamic acid derivatives may have dual inhibitory activities against both pfLDH and pfMDH. It is presumed that dual LDH/MDH inhibitors would have enhanced potential as antimalarial drugs.
Sigma-Aldrich
Oxamic acid, ≥98%