Merck
  • Defective signaling through plexin-A1 compromises the development of the peripheral olfactory system and neuroendocrine reproductive axis in mice.
Defective signaling through plexin-A1 compromises the development of the peripheral olfactory system and neuroendocrine reproductive axis in mice.
Human molecular genetics (2017-03-24)
Séverine Marcos, Carine Monnier, Xavier Rovira, Corinne Fouveaut, Nelly Pitteloud, Fabrice Ango, Catherine Dodé, Jean-Pierre Hardelin
ABSTRACT
The olfacto-genital syndrome (Kallmann syndrome) associates congenital hypogonadism due to gonadotropin-releasing hormone (GnRH) deficiency and anosmia. This is a genetically heterogeneous developmental disease with various modes of transmission, including oligogenic inheritance. Previous reports have involved defective cell signaling by semaphorin-3A in the disease pathogenesis. Here, we report that the embryonic phenotype of Plxna1-/- mutant mice lacking plexin-A1 (a major receptor of class 3 semaphorins), though not fully penetrant, resembles that of Kallmann syndrome fetuses. Pathohistological analysis indeed showed a strongly abnormal development of the peripheral olfactory system and defective embryonic migration of the neuroendocrine GnRH cells to the hypothalamic brain region in some of the mutant mice, which resulted in reduced fertility in adult males. We thus screened 250 patients for the presence of mutations in PLXNA1, and identified different nonsynonymous mutations (p.V349L, p.V437L, p.R528W, p.H684Y, p.G720E, p.R740H, p.R813H, p.R840Q, p.A854T, p.R897H, p.L1464V, p.K1618T, p.C1744F), all at heterozygous state, in 15 patients. Most of these mutations are predicted to affect plexin-A1 stability or signaling activity based on predictive algorithms and a structural model of the protein. Moreover, in vitro experiments allowed us to show the existence of deleterious effects of eight mutations (including a transcript splicing defect), none of which are expected to result in a complete loss of protein synthesis, targeting, or signaling activity, though. Our findings indicate that signaling insufficiency through plexin-A1 can contribute to the pathogenesis of Kallmann syndrome, and further substantiate the oligogenic pattern of inheritance in this developmental disorder.
Sigma-Aldrich
Anti-Peripherin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Plexin-A1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Brain lipid binding protein Antibody, from rabbit, purified by affinity chromatography