Merck
  • Synthesis and antimicrobial activity of some novel phenyl and benzimidazole substituted benzyl ethers.
Synthesis and antimicrobial activity of some novel phenyl and benzimidazole substituted benzyl ethers.
Bioorganic & medicinal chemistry letters (2007-02-10)
Ozden Ozel Güven, Taner Erdoğan, Hakan Göker, Sulhiye Yildiz
ABSTRACT
In this study, a series of novel phenyl- and benzimidazole-substituted benzyl ethers were synthesized and evaluated for antibacterial and antifungal activities against Staphylococcus aureus, Methicillin-resistant S. aureus (MRSA), Escherichia coli, Candida albicans, and Candida krusei. Compound 6g exhibited the most potent antibacterial activity with lowest MIC values of 3.12 and 6.25 microg/mL against S. aureus and MRSA, respectively.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard