Merck
Total Synthesis of Phorbazole B.
Molecules (Basel, Switzerland) (2020-10-25)
Yngve Guttormsen, Magnus E Fairhurst, Sunil K Pandey, Johan Isaksson, Bengt Erik Haug, Annette Bayer
ABSTRACT
Phorbazoles are polychlorinated heterocyclic secondary metabolites isolated from a marine sponge and several of these natural products have shown inhibitory activity against cancer cells. In this work, a synthesis of the trichlorinated phorbazole B using late stage electrophilic chlorination was developed. The synthesis relied on the use of an oxazole precursor, which was protected with an iodine in the reactive 4-position, followed by complete chlorination of all pyrrole positions. Attempts to prepare phorbazole A and C, which contain a 3,4-dichlorinated pyrrole, were unsuccessful as the desired chlorination pattern on the pyrrole could not be obtained. The identities of the dichlorinated intermediates and products were determined using NMR techniques including NOESY/ROESY, 1,1-ADEQUATE and high-resolution CLIP-HSQMBC.
Sigma-Aldrich
Sodium permanganate solution, 40 wt. % in H2O
Pricing and availability is not currently available.