Merck
  • Synthesis and anti-inflammatory activity of 3-(4'-geranyloxy-3'-methoxyphenyl)-2-trans propenoic acid and its ester derivatives.
Synthesis and anti-inflammatory activity of 3-(4'-geranyloxy-3'-methoxyphenyl)-2-trans propenoic acid and its ester derivatives.
Bioorganic & medicinal chemistry letters (2007-09-05)
Francesco Epifano, Salvatore Genovese, Silvio Sosa, Aurelia Tubaro, Massimo Curini
ABSTRACT
Different esters of 3-(4'-geranyloxy-3'-methoxyphenyl)-2-trans propenoic acid (1), an anti-inflammatory principle of Acronychia baueri Schott (Rutaceae), were synthesized. Their topical anti-inflammatory activity was evaluated using the Croton oil ear test in mice as a model of acute inflammation. The activity of the paracetamol, guaiacol and hydroquinone esters of (1) was higher than that of the parent compound, being similar to that exerted by indomethacin, used as reference drug.
Product Description
Supelco
trans-Ferulic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Vanillic acid, 97%
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)