Merck
  • Synthesis and TNF expression inhibitory properties of new thalidomide analogues derived via Heck cross coupling.
Synthesis and TNF expression inhibitory properties of new thalidomide analogues derived via Heck cross coupling.
Bioorganic & medicinal chemistry letters (2007-09-14)
Scott G Stewart, Daniel Spagnolo, Marta E Polomska, Melvin Sin, Mahdad Karimi, Lawrence J Abraham
ABSTRACT
A library of new thalidomide analogues containing an olefin functionality were synthesised using a Heck cross coupling reaction from their aryl halogenated precursor. All analogues were tested for their ability to inhibit the synthesis of the proinflammatory cytokine Tumour Necrosis Factor (TNF). Compounds 22, 29, 33 and 37 were the most effective in this assay inhibiting TNF expression 50%, 69%, 52% and 50%, respectively.
Sigma-Aldrich
(±)-Thalidomide, ≥98%, powder