Merck
A Leptin-Mediated Neural Mechanism Linking Breathing to Metabolism.
Cell reports (2020-11-12)
Jeehaeh Do, Zheng Chang, Gabriella Sekerková, Donald R McCrimmon, Marco Martina
ABSTRACT
Breathing is coupled to metabolism. Leptin, a peptide mainly secreted in proportion to adipose tissue mass, increases energy expenditure with a parallel increase in breathing. We demonstrate that optogenetic activation of LepRb neurons in the nucleus of the solitary tract (NTS) mimics the respiratory stimulation after systemic leptin administration. We show that leptin activates the sodium leak channel (NALCN), thereby depolarizing a subset of glutamatergic (VGluT2) LepRb NTS neurons expressing galanin. Mice with selective deletion of NALCN in LepRb neurons have increased breathing irregularity and central apneas. On a high-fat diet, these mice gain weight with an associated depression of minute ventilation and tidal volume, which are not detected in control littermates. Anatomical mapping reveals LepRb NTS-originating glutamatergic axon terminals in a brainstem inspiratory premotor region (rVRG) and dorsomedial hypothalamus. These findings directly link a defined subset of NTS LepRb cells to the matching of ventilation to energy balance.
Sigma-Aldrich
Anti-c-Fos Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-VGluT2 Antibody, serum, from guinea pig