Merck
Synthesis, antimicrobial, and QSAR studies of substituted benzamides.
Bioorganic & medicinal chemistry (2007-04-13)
Anil Kumar, Balasubramanian Narasimhan, Devinder Kumar
ABSTRACT
A series of new substituted benzamides were synthesized and tested in vitro for their antibacterial activity against Gram-positive and Gram-negative bacteria and as well for antifungal activity. The compounds 8i and 9 showed better activity among the different benzamides synthesized. The structural characteristics governing antibacterial activities of substituted benzamides were studied using QSAR methodology. The results showed that the antimicrobial activity could be modeled using the topological descriptors, molecular connectivity indices (2chi(v) and 2chi) and Kiers shape index (kappaalpha1). The low residual activity and high cross-validated r2 values (r(cv)2) observed indicated the predictive ability of the developed QSAR models.
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Oxyclozanide, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)