Merck
  • Glycogen synthase kinase-3 inhibition rescues sex-dependent contextual fear memory deficit in human immunodeficiency virus-1 transgenic mice.
Glycogen synthase kinase-3 inhibition rescues sex-dependent contextual fear memory deficit in human immunodeficiency virus-1 transgenic mice.
British journal of pharmacology (2020-10-21)
Shamsudheen Moidunny, Michael A Benneyworth, David J Titus, Eleonore Beurel, Udhghatri Kolli, Joyce Meints, Richa Jalodia, Sundaram Ramakrishnan, Coleen M Atkins, Sabita Roy, Shamsudheen Moidunny, Michael A Benneyworth, David J Titus, Eleonore Beurel, Udhghatri Kolli, Joyce Meints, Richa Jalodia, Sundaram Ramakrishnan, Coleen M Atkins, Sabita Roy
ABSTRACT
A significant number of HIV-1 patients on antiretroviral therapy develop HIV-associated neurocognitive disorders (HAND). Evidence indicate that biological sex may regulate HAND pathogenesis, but the mechanisms remain unknown. We investigated synaptic mechanisms associated with sex differences in HAND, using the HIV-1-transgenic 26 (Tg26) mouse model. Contextual- and cue-dependent memories of male and female Tg26 mice and littermate wild type mice were assessed in a fear conditioning paradigm. Hippocampal electrophysiology, immunohistochemistry, western blot, qRT-PCR and ELISA techniques were used to investigate cellular, synaptic and molecular impairments. Cue-dependent memory was unaltered in male and female Tg26 mice, when compared to wild type mice. Male, but not female, Tg26 mice showed deficits in contextual fear memory. Consistently, only male Tg26 mice showed depressed hippocampal basal synaptic transmission and impaired LTP induction in area CA1. These deficits in male Tg26 mice were independent of hippocampal neuronal loss and microglial activation but were associated with increased HIV-1 long terminal repeat mRNA expression, reduced hippocampal synapsin-1 protein, reduced BDNF mRNA and protein, reduced AMPA glutamate receptor (GluA1) phosphorylation levels and increased glycogen synthase kinase 3 (GSK3) activity. Importantly, selective GSK3 inhibition using 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione increased levels of synapsin-1, BDNF and phosphorylated-GluA1 proteins, restored hippocampal basal synaptic transmission and LTP, and improved contextual fear memory in male Tg26 mice. Sex-dependent impairments in contextual fear memory and synaptic plasticity in Tg26 mice are associated with increased GSK3 activity. This implicates GSK3 inhibition as a potential therapeutic strategy to improve cognition in HIV-1 patients.
Sigma-Aldrich
RIPA Buffer
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
TDZD-8, ≥98% (HPLC), needles
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp PCR ReadyMix, Ready-to-use 2X PCR Master Mix with Loading Dye
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GSK3 Antibody, clone 4G-1E, clone 4G-1E, Upstate®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-phospho-GluR1 (Ser831) Antibody, clone N453, rabbit monoclonal, culture supernatant, clone N453, Upstate®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SY38, clone SY38, Chemicon®, from mouse
Pricing and availability is not currently available.