Merck
  • Antiprotozoal and antimicrobial activities of O-alkylated and formylated acylphloroglucinols.
Antiprotozoal and antimicrobial activities of O-alkylated and formylated acylphloroglucinols.
Bioorganic & medicinal chemistry (2006-10-31)
Sandip B Bharate, Shabana I Khan, Nafees A M Yunus, Siddheshwar K Chauthe, Melissa R Jacob, Babu L Tekwani, Ikhlas A Khan, Inder Pal Singh
ABSTRACT
In the present article, we examined the antileishmanial, antimalarial, antibacterial, and antifungal activities of several newly synthesized O-alkylated phloroglucinol compounds (11-19) which are analogues of the naturally occurring antimalarial compound 1. Analogues 12 and 16 exhibited antileishmanial activity against, Leishmania donovani promastigotes with IC(50)s of 5.3 and 4.2microg/mL, respectively. Naturally occurring monomeric formylated acylphloroglucinol compounds, grandinol (2), jensenone (3), and their analogues (29-37), were also synthesized and evaluated for antileishmanial, antimalarial, antibacterial, and antifungal activities. Amongst these, both grandinol and jensenone showed mild to moderate antibacterial, antifungal, and antileishmanial activities. Jensenone (3) was effective against Candida albicans with an IC(50) of 5.5microg/mL but was ineffective against Cryptococcus neoformans and methicillin-resistant Staphylococcus aureus. Among the analogues, 34 was the most active against C. albicans and C. neoformans with IC(50)s of 2.0 and 2.5microg/mL, respectively, and was fungicidal toward Candida albicans.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard