Merck
  • Suppression of Netrin-1 attenuates angiotension II-induced cardiac remodeling through the PKC/MAPK signaling pathway.
Suppression of Netrin-1 attenuates angiotension II-induced cardiac remodeling through the PKC/MAPK signaling pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-07-21)
Gaojun Wu, Zhengxian Wang, Peiren Shan, Shanjun Huang, Shuang Lin, Weijian Huang, Zhouqing Huang
ABSTRACT
Myocardial remodeling caused by angiotensin II (Ang II) is essential for the pathological process of heart failure. Netrin-1, which is an axonal guidance cue, has been shown to be involved in the inflammatory response, tumorigenesis, and angiogenesis in non-neuronal tissues. However, the role of Netrin-1 in cardiac remodeling has not been fully elucidated. The rat cardiomyocyte cell line H9c2 and primary neonatal rat cardiomyocytes were treated with Ang II. Cells were transfected with siRNA to silence Netrin-1 expression. Real-time polymerase chain reaction and Western blot analysis were used to detect the markers for fibrosis, apoptosis, and hypertrophy in cardiomyocytes. An Annexin V-EGFP/PI cell apoptosis detection kit was used to measure the level of apoptosis caused by angiotensin II. We found that Netrin-1 expression was upregulated in the H9c2 cells and the neonatal rat cardiomyocytes stimulated by Ang II. The increased Netrin-1 expression was decreased by valsartan to block AT1R. Importantly, the application of Netrin-1 siRNA significantly alleviated the degrees of myocardial hypertrophy, fibrosis (reflected by Myhc, collagen I, and TGF-β) and apoptosis (reflected by the level of Caspase 3, Bax, and Bcl-2) induced by Ang II. In addition, the silencing of Netrin-1 substantially decreased the phosphorylation of PKCα, JNK, and P38. We treated H9c2 cells with LY317615, SP600125, and SB203580, inhibitors of PKCα, JNK, and P38, respectively, thereby resulting in a substantial decrease in hypertrophy, fibrosis, and apoptosis. Ang II produces cardiac hypertrophy, fibrosis, and apoptosis through the upregulation of Netrin-1 and the activation of the AT1R/PKCα/MAPK (JNK, P38) pathway. Suppression of Netrin-1 can relieve Ang II-induced cardiac remodeling via inhibition of the PKCα/MAPK (JNK and P38) signaling pathway. Thus, Netrin-1 may be a novel therapeutic target for Ang II-mediated cardiac remodeling.
Sigma-Aldrich
Valsartan, ≥98% (HPLC)