Merck
Neoglycosylated Collagen: Effect on Neuroblastoma F-11 Cell Lines.
Molecules (Basel, Switzerland) (2020-09-27)
Antonella Sgambato, Valentina Pastori, Laura Russo, Simone Vesentini, Marzia Lecchi, Laura Cipolla
ABSTRACT
The regeneration of the nervous system is a challenging task. Currently, regenerative medicine approaches that exploit nature-inspired cues are being studied and hold great promise. The possibility to use protein-based matrices functionalized with small oligo- and monosaccharides is of interest since these can be finely tuned to better mimic the native environment. Collagen has been selected as a promising material that has the potential to be further tailored to incorporate carbohydrates in order to drive cell behavior towards neuroregeneration. Indeed, the grafting of carbohydrates to collagen 2D matrices is proved to enhance its biological significance. In the present study, collagen 2D matrices were grafted with different carbohydrate epitopes, and their potential to drive F-11 neuroblastoma cells towards neuronal differentiation was evaluated. Collagen functionalized with α-glucosides was able to differentiate neuroblastoma cells into functional neurons, while sialyl α-(2→6)-galactosides stimulated cell proliferation.
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase