Merck
Designing better coumarin-based fluorogenic substrates for PTP1B.
Bioorganic & medicinal chemistry letters (2008-04-30)
Christopher P Holmes, Natalie Macher, J Russell Grove, Larry Jang, Jennifer D Irvine
ABSTRACT
As part of a program to develop better PTP1B fluorogenic substrates that more closely mimic the functionality found in the natural substrate, we have prepared and evaluated nine novel analogs of 4-methylumbelliferone phosphate (MUP) with a variety of additional groups occupying the second phosphate binding pocket.
Sigma-Aldrich
4-Methylumbelliferyl phosphate (4-MUP) Liquid Substrate System, liquid
Sigma-Aldrich
4-Methylumbelliferyl phosphate, phosphatase substrate