Merck
  • Synthesis and cytotoxic activity of some novel polycyclic gamma-butyrolactones.
Synthesis and cytotoxic activity of some novel polycyclic gamma-butyrolactones.
Bioorganic & medicinal chemistry letters (2008-08-19)
M P S Ishar, Tilak Raj, Satyam Kumar Agrawal, A K Saxena, Lakhwinder Singh, Rajinder Singh, Surinderjit Singh Bhella
ABSTRACT
Baeyer-Villiger oxidation of 5-aryl-7,11,11-trimethyltricyclo[5.4.0.0(3,6)]-undec-1-en-4-ones 4a-h by H(2)O(2) and formic acid in methanol yields mixtures of 3b,7,7-trimethyl-3-phenyl-3,3a,3b,4,5,6,7,8a-octahydro-1H-indeno-[1,2-c]furan-1-ones 8a-h and 3b,7,7-trimethyl-3-phenyl-3,3a,3b,4,5,6,7,8a-octahydro-1H-indeno-[1,2-c]furan-2-ones 9a-h in high yields. The obtained butyrolactones 8a-h display cytotoxic activity against a number of human cancer cells.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture