Merck
  • Aroyl hydrazones of 2-phenylindole-3-carbaldehydes as novel antimitotic agents.
Aroyl hydrazones of 2-phenylindole-3-carbaldehydes as novel antimitotic agents.
Bioorganic & medicinal chemistry (2008-06-03)
Susanne Vogel, Doris Kaufmann, Michaela Pojarová, Christine Müller, Tobias Pfaller, Sybille Kühne, Patrick J Bednarski, Erwin von Angerer
ABSTRACT
Cell cycle arrest of malignant cells is an important option for cancer treatment. In this study, we modified the structure of antimitotic 2-phenylindole-3-carbaldehydes by condensation with hydrazides of various benzoic and pyridine carboxylic acids. The resulting hydrazones inhibited the growth of MDA-MB 231 and MCF-7 breast cancer cells with IC(50) values of 20-30 nM for the most potent derivatives. These 2-phenylindole derivatives also exerted an inhibitory effect on the growth of both proliferating and resting U-373 MG glioblastoma cells. Though the hydrazones exhibited similar structure-activity relationships as the aldehydes, they did not inhibit tubulin polymerization as the aldehydes but were capable of blocking the cell cycle in G(2)/M phase. The cell cycle arrest was accompanied by apoptosis as demonstrated by the activation of caspase-3. Since these 2-phenylindole-based hydrazones display no structural similarity with other antitumor drugs they are interesting candidates for further development.
Product Description
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Colchicine, suitable for plant cell culture, BioReagent, ≥95% (HPLC)