Merck
  • Cytosolic, but not matrix, calcium is essential for adjustment of mitochondrial pyruvate supply.
Cytosolic, but not matrix, calcium is essential for adjustment of mitochondrial pyruvate supply.
The Journal of biological chemistry (2020-02-26)
Marten Szibor, Zemfira Gizatullina, Timur Gainutdinov, Thomas Endres, Grazyna Debska-Vielhaber, Matthias Kunz, Niki Karavasili, Kerstin Hallmann, Frank Schreiber, Alexandra Bamberger, Michael Schwarzer, Torsten Doenst, Hans-Jochen Heinze, Volkmar Lessmann, Stefan Vielhaber, Wolfram S Kunz, Frank N Gellerich
ABSTRACT
Mitochondrial oxidative phosphorylation (OXPHOS) and cellular workload are tightly balanced by the key cellular regulator, calcium (Ca2+). Current models assume that cytosolic Ca2+ regulates workload and that mitochondrial Ca2+ uptake precedes activation of matrix dehydrogenases, thereby matching OXPHOS substrate supply to ATP demand. Surprisingly, knockout (KO) of the mitochondrial Ca2+ uniporter (MCU) in mice results in only minimal phenotypic changes and does not alter OXPHOS. This implies that adaptive activation of mitochondrial dehydrogenases by intramitochondrial Ca2+ cannot be the exclusive mechanism for OXPHOS control. We hypothesized that cytosolic Ca2+, but not mitochondrial matrix Ca2+, may adapt OXPHOS to workload by adjusting the rate of pyruvate supply from the cytosol to the mitochondria. Here, we studied the role of malate-aspartate shuttle (MAS)-dependent substrate supply in OXPHOS responses to changing Ca2+ concentrations in isolated brain and heart mitochondria, synaptosomes, fibroblasts, and thymocytes from WT and MCU KO mice and the isolated working rat heart. Our results indicate that extramitochondrial Ca2+ controls up to 85% of maximal pyruvate-driven OXPHOS rates, mediated by the activity of the complete MAS, and that intramitochondrial Ca2+ accounts for the remaining 15%. Of note, the complete MAS, as applied here, included besides its classical NADH oxidation reaction the generation of cytosolic pyruvate. Part of this largely neglected mechanism has previously been described as the "mitochondrial gas pedal." Its implementation into OXPHOS control models integrates seemingly contradictory results and warrants a critical reappraisal of metabolic control mechanisms in health and disease.
Product Description
Roche
Penicillin-Streptomycin, suitable for cell culture, lyophilized (clear, colorless solution after reconstitution), non-sterile; 0.2 μm filtered, sufficient for 20 mL
Pricing and availability is not currently available.
Sigma-Aldrich
O-(Carboxymethyl)hydroxylamine hemihydrochloride, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ~98%, pkg of 100 mg
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
Pricing and availability is not currently available.
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium azide, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Tylosin solution, 8 mg/mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Ruthenium Red, Technical grade
Pricing and availability is not currently available.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium L-lactate, ~98%
Pricing and availability is not currently available.
Millipore
BCA Protein Assay Kit, The BCA protein assay is a simple and reliable protein quantification method.
Pricing and availability is not currently available.
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamic acid monosodium salt monohydrate, ≥98.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
Rotenone, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Pricing and availability is not currently available.