Merck
  • Bacillus cereus non-haemolytic enterotoxin activates the NLRP3 inflammasome.
Bacillus cereus non-haemolytic enterotoxin activates the NLRP3 inflammasome.
Nature communications (2020-02-08)
Daniel Fox, Anukriti Mathur, Yansong Xue, Yunqi Liu, Wei Hong Tan, Shouya Feng, Abhimanu Pandey, Chinh Ngo, Jenni A Hayward, Ines I Atmosukarto, Jason D Price, Matthew D Johnson, Nadja Jessberger, Avril A B Robertson, Gaetan Burgio, David C Tscharke, Edward M Fox, Denisse L Leyton, Nadeem O Kaakoush, Erwin Märtlbauer, Stephen H Leppla, Si Ming Man
ABSTRACT
Inflammasomes are important for host defence against pathogens and homeostasis with commensal microbes. Here, we show non-haemolytic enterotoxin (NHE) from the neglected human foodborne pathogen Bacillus cereus is an activator of the NLRP3 inflammasome and pyroptosis. NHE is a non-redundant toxin to haemolysin BL (HBL) despite having a similar mechanism of action. Via a putative transmembrane region, subunit C of NHE initiates binding to the plasma membrane, leading to the recruitment of subunit B and subunit A, thus forming a tripartite lytic pore that is permissive to efflux of potassium. NHE mediates killing of cells from multiple lineages and hosts, highlighting a versatile functional repertoire in different host species. These data indicate that NHE and HBL operate synergistically to induce inflammation and show that multiple virulence factors from the same pathogen with conserved function and mechanism of action can be exploited for sensing by a single inflammasome.
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, ≥98% (HPLC), powder
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Roche
DOTAP Liposomal Transfection Reagent, >99% (TLC), liquid, suitable for transfection
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture