Merck
  • Hydrofluoric acid treatment of titanium surfaces enhances the proliferation of human gingival fibroblasts.
Hydrofluoric acid treatment of titanium surfaces enhances the proliferation of human gingival fibroblasts.
Journal of tissue engineering (2019-02-26)
Maria H Pham, Håvard J Haugen, Alessandra Rinna, Jan Eirik Ellingsen, Janne E Reseland
ABSTRACT
The attachment of implants relies on bone and soft tissue biocompatibility. The aim of this article is to investigate the effect of fluoride-modified metallic titanium (Ti) surfaces (Ti-F) on proliferation and differentiation of human gingival fibroblasts. Human gingival fibroblast cells were exposed to hydrofluoric acid-modified Ti coins (Ti-F) for 1, 3, 7, 14 and 21 days, and untreated coins were used as controls. A five- to six-fold increase in the proliferation of human gingival fibroblasts on Ti-F compared to Ti surfaces was observed. Enhanced gene expression of interleukin-6 and osteoprotegerin was found at 7 days. Increased levels of sclerostin, interleukin-6 and osteoprotegerin in the media from human gingival fibroblasts cultured on Ti-F coins were found compared to controls. Our results confirm that hydrofluoric acid-modified surface may indirectly enhance the firm attachment of implant surface to junction epithelium, soft tissue epithelium, which would give protection for underlying osseous structures making osseointegration of the dental implant possible.
Product Description
Millipore
MILLIPLEX® Human Bone Panel, Configurable Human Bone 13-Plex Panel
Millipore
MILLIPLEX® Human RANKL Singleplex, Human RANKL Singleplex