Merck
  • Leukemia inhibitory factor deficiency modulates the immune response and limits autoimmune demyelination: a new role for neurotrophic cytokines in neuroinflammation.
Leukemia inhibitory factor deficiency modulates the immune response and limits autoimmune demyelination: a new role for neurotrophic cytokines in neuroinflammation.
Journal of immunology (Baltimore, Md. : 1950) (2008-02-06)
Ralf A Linker, Niels Kruse, Stephanie Israel, Tao Wei, Silvia Seubert, Anja Hombach, Bettina Holtmann, Fred Luhder, Richard M Ransohoff, Michael Sendtner, Ralf Gold
ABSTRACT
The neurotrophic cytokines ciliary neurotrophic factor and leukemia inhibitory factor (LIF) play a key role in neuronal and oligodendrocyte survival and as protective factors in neuroinflammation. To further elucidate the potential of endogenous LIF in modulating neuroinflammation, we studied myelin oligodendrocyte glycoprotein-induced experimental autoimmune encephalomyelitis in LIF knockout mice (LIF(-/-) mice). In the late phase of active myelin oligodendrocyte glycoprotein-induced experimental autoimmune encephalomyelitis, LIF(-/-) mice exhibited a markedly milder disease course. The inflammatory infiltrate in LIF(-/-) mice was characterized by an increase in neutrophilic granulocytes early and fewer infiltrating macrophages associated with less demyelination later in the disease. In good correlation with an effect of endogenous LIF on the immune response, we found an Ag-specific T cell-priming defect with impaired IFN-gamma production in LIF(-/-) mice. On the molecular level, the altered recruitment of inflammatory cells is associated with distinct patterns of chemokine production in LIF(-/-) mice with an increase of CXCL1 early and a decrease of CCL2, CCL3, and CXCL10 later in the disease. These data reveal that endogenous LIF is an immunologically active molecule in neuroinflammation. This establishes a link between LIF and the immune system which was not observed in the ciliary neurotrophic factor knockout mouse.
Sigma-Aldrich
Leukocyte Naphthol AS-D Chloroacetate (Specific Esterase) Kit
Pricing and availability is not currently available.