Merck
  • COX, LOX and platelet aggregation inhibitory properties of Lauraceae neolignans.
COX, LOX and platelet aggregation inhibitory properties of Lauraceae neolignans.
Bioorganic & medicinal chemistry letters (2009-11-03)
Ericsson David Coy, Luis Enrique Cuca, Michael Sefkow
ABSTRACT
The anti-inflammatory potential of 26 neolignans (14 of the bicyclooctane-type and 12 of the benzofuran-type), isolated from three Lauraceae species (Pleurothyrium cinereum, Ocotea macrophylla and Nectandra amazonum), was evaluated in vitro through inhibition of COX-1, COX-2, 5-LOX and agonist-induced aggregation of rabbit platelets. Benzofuran neolignans were found to be selective COX-2 inhibitors, whereas bicyclooctane neolignans inhibit selectively the PAF-action as well as COX-1 and 5-LOX. The neolignan 9-nor-7,8-dehydro-isolicarin B 15 and cinerin C 7 were found to be the most potent COX-2 inhibitor and PAF-antagonist, respectively. Nectamazin C 10 exhibited dual 5-LOX/COX-2 inhibition.
Product Description
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard