Merck
  • Circulating Exosomal miR-96 as a Novel Biomarker for Radioresistant Non-Small-Cell Lung Cancer.
Circulating Exosomal miR-96 as a Novel Biomarker for Radioresistant Non-Small-Cell Lung Cancer.
Journal of oncology (2021-03-18)
Qin Zheng, Huaiyin Ding, Lixue Wang, Yu Yan, Yuan Wan, Yongxiang Yi, Liang Tao, Chuandong Zhu
ABSTRACT
Patients with non-small-cell lung cancer (NSCLC) frequently develop radioresistance, resulting in poor response to radiation and unfavourable prognosis. Early detection of radioresistance hence can guide the adjustment of treatment regimens in time. Exosomes are lipid bilayer-enclosed vesicles with sub-micrometer size that are released by various cells. Exosomes contain a tissue-specific signature wherein a variety of proteins and nucleic acids are selectively packaged. Growing evidence shows exosomes are involved in cancer pathophysiology and exosomes as the latest addition to the liquid biopsy portfolio have been used in cancer diagnosis. Compared to cell free RNA, exosomal lipid envelope can effectively protect RNA cargo against degradation. Therefore, exosomes may hold great promise for the identification of radioresistance. Here, we report six plasma exosomal miRNAs could be used to distinguish radioresistant NSCLC patients from radiosensitive NSCLC patients and to evaluate the prognosis of NSCLC. Samples were obtained from 52 NSCLC patients with or without radioresistance and 45 age-matched healthy volunteers. Exosomes in 1 ml plasma were isolated followed by extraction of small RNA. The expression levels of miRNAs were determined by quantitative real-time PCR. Potential miRNA markers were further evaluated in additional 52 NSCLC patients. We found exosomal miR-1246 and miR-96 are significantly overexpressed in NSCLC patients. Moreover, exosomal miR-96 in patients with radioresistant NSCLC is significantly higher than that of controls. Exosomal miR-96 also demonstrates a significant correlation with vascular invasion and poor overall survival. Altogether, our results indicate that exosomal miR-96 could be a non-invasive diagnostic and prognostic marker of radioresistant NSCLC.
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Diluent C for General Membrane Labeling, Distributed for Phanos Technologies