Merck
  • Development and biochemical and immunological characterization of early passage and immortalized bovine intestinal epithelial cell lines from the ileum of a young calf.
Development and biochemical and immunological characterization of early passage and immortalized bovine intestinal epithelial cell lines from the ileum of a young calf.
Cytotechnology (2019-01-03)
Pratik Katwal, Milton Thomas, Tirth Uprety, Michael B Hildreth, Radhey S Kaushik
ABSTRACT
The intestinal epithelium is a major site of interaction with pathogens. In bovine intestinal epithelial cells (BIECs), Toll-like receptors (TLRs) play an important role in innate immune responses against enteric pathogens. This study is aimed at establishing a stable bovine intestinal epithelial cell line that can be maintained by a continuous passage so that studies on innate immune responses against various enteric pathogens can be performed. The main goal was to establish pure cultures of primary and immortalized bovine intestinal epithelial cells from the ileum and then characterize them biochemically and immunologically. Mixed epithelial and fibroblast bovine ileal intestinal cultures were first established from a 2-day old calf. Limiting dilution method was used to obtain a clone of epithelial cells which was characterized using immunocytochemistry (ICC). The selected clone BIEC-c4 was cytokeratin positive and expressed low levels of vimentin, confirming the epithelial cell phenotype. Early passage BIEC-c4 cells were transfected with either simian virus 40 (SV40) large T antigen or human telomerase reverse transcriptase (hTERT), or human papillomavirus (HPV) type 16E6/E7 genes to establish three immortalized BIEC cell lines. The expression of SV40, hTERT and HPV E6/E7 genes in immortalized BIECs was confirmed by a polymerase chain reaction (PCR). Immunocytochemistry and immunofluorescence assays also confirmed the expression of SV40, hTERT and HPV E6 proteins. The immortalized BIECs were cytokeratin positive and all except HPV-BIECs expressed low levels of vimentin. A growth kinetics study indicated that there were no significant differences in the doubling time of immortalized BIECs as compared to early passage BIEC-c4 cells. All four BIEC types expressed TLR 1-10 genes, with TLR 3 and 4 showing higher expression across all cell types. These newly established early passage and immortalized BIEC cell lines should serve as a good model for studying infectivity, pathogenesis and innate immune responses against enteric pathogens.
Sigma-Aldrich
IgM, Lambda from murine myeloma, clone MOPC 104E, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG2a, Kappa from murine myeloma, clone UPC 10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone VIM-13.2, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Desmin antibody produced in mouse, clone DE-U-10, ascites fluid