Merck
  • Besifloxacin, a novel fluoroquinolone, has broad-spectrum in vitro activity against aerobic and anaerobic bacteria.
Besifloxacin, a novel fluoroquinolone, has broad-spectrum in vitro activity against aerobic and anaerobic bacteria.
Antimicrobial agents and chemotherapy (2009-06-10)
Wolfgang Haas, Chris M Pillar, Gary E Zurenko, Jacqueline C Lee, Lynne S Brunner, Timothy W Morris
ABSTRACT
The antibacterial spectrum of besifloxacin, a novel fluoroquinolone recently approved for treatment of ocular infections, was studied using 2,690 clinical isolates representing 40 species. Overall, besifloxacin was the most potent agent tested against gram-positive pathogens and anaerobes and was generally equivalent to comparator fluoroquinolones in activity against most gram-negative pathogens. Besifloxacin demonstrated potent, broad-spectrum activity, which was particularly notable against gram-positive and gram-negative isolates that were resistant to other fluoroquinolones and classes of antibacterial agents.
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)