Merck
  • Purified Smc5/6 Complex Exhibits DNA Substrate Recognition and Compaction.
Purified Smc5/6 Complex Exhibits DNA Substrate Recognition and Compaction.
Molecular cell (2020-12-11)
Pilar Gutierrez-Escribano, Silvia Hormeño, Julene Madariaga-Marcos, Roger Solé-Soler, Francis J O'Reilly, Kyle Morris, Clara Aicart-Ramos, Ricardo Aramayo, Alex Montoya, Holger Kramer, Juri Rappsilber, Jordi Torres-Rosell, Fernando Moreno-Herrero, Luis Aragon
ABSTRACT
Eukaryotic SMC complexes, cohesin, condensin, and Smc5/6, use ATP hydrolysis to power a plethora of functions requiring organization and restructuring of eukaryotic chromosomes in interphase and during mitosis. The Smc5/6 mechanism of action and its activity on DNA are largely unknown. Here we purified the budding yeast Smc5/6 holocomplex and characterized its core biochemical and biophysical activities. Purified Smc5/6 exhibits DNA-dependent ATP hydrolysis and SUMO E3 ligase activity. We show that Smc5/6 binds DNA topologically with affinity for supercoiled and catenated DNA templates. Employing single-molecule assays to analyze the functional and dynamic characteristics of Smc5/6 bound to DNA, we show that Smc5/6 locks DNA plectonemes and can compact DNA in an ATP-dependent manner. These results demonstrate that the Smc5/6 complex recognizes DNA tertiary structures involving juxtaposed helices and might modulate DNA topology by plectoneme stabilization and local compaction.
Sigma-Aldrich
L-Lactate Dehydrogenase (LDHA), from human, recombinant, expressed in E. coli, aqueous solution
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Phospho(enol)pyruvic acid monopotassium salt, ≥97% (enzymatic)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial