Merck
  • Inhibition of two-pore channels in antigen-presenting cells promotes the expansion of TNFR2-expressing CD4+Foxp3+ regulatory T cells.
Inhibition of two-pore channels in antigen-presenting cells promotes the expansion of TNFR2-expressing CD4+Foxp3+ regulatory T cells.
Science advances (2020-10-02)
Tianzhen He, De Yang, Xiao-Qing Li, Mengmeng Jiang, Md Sahidul Islam, Shaokui Chen, Yibo Chen, Yang Yang, Chon-Kit Chou, Anna L Trivett, Joost J Oppenheim, Xin Chen
ABSTRACT
CD4+Foxp3+ regulatory T cells (Tregs) are pivotal for the inhibition of autoimmune inflammatory responses. One way to therapeutically harness the immunosuppressive actions of Tregs is to stimulate the proliferative expansion of TNFR2-expressing CD4+Foxp3+ Tregs via transmembrane TNF (tmTNF). Here, we report that two-pore channel (TPC) inhibitors markedly enhance tmTNF expression on antigen-presenting cells. Furthermore, injection of TPC inhibitors including tetrandrine, or TPC-specific siRNAs in mice, increases the number of Tregs in a tmTNF/TNFR2-dependent manner. In a mouse colitis model, inhibition of TPCs by tetrandrine markedly attenuates colon inflammation by expansion of Tregs Mechanistically, we show that TPC inhibitors enhance tmTNF levels by disrupting surface expression of TNF-α-converting enzyme by regulating vesicle trafficking. These results suggest that the therapeutic potential of TPC inhibitors is mediated by expansion of TNFR2-expressing Tregs and elucidate the basis of clinical use in the treatment of autoimmune and other inflammatory diseases.
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Gabapentin, solid
Sigma-Aldrich
Nimodipine
Sigma-Aldrich
Nilotinib
Sigma-Aldrich
ω-Agatoxin IVA, ≥90% (HPLC)
Sigma-Aldrich
(S)-(−)-N-(Trifluoroacetyl)pyrrolidine-2-carbonyl chloride solution, 0.1 M in methylene chloride
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder