Merck
  • Clinically relevant mutations in core metabolic genes confer antibiotic resistance.
Clinically relevant mutations in core metabolic genes confer antibiotic resistance.
Science (New York, N.Y.) (2021-02-20)
Allison J Lopatkin, Sarah C Bening, Abigail L Manson, Jonathan M Stokes, Michael A Kohanski, Ahmed H Badran, Ashlee M Earl, Nicole J Cheney, Jason H Yang, James J Collins
ABSTRACT
Although metabolism plays an active role in antibiotic lethality, antibiotic resistance is generally associated with drug target modification, enzymatic inactivation, and/or transport rather than metabolic processes. Evolution experiments of Escherichia coli rely on growth-dependent selection, which may provide a limited view of the antibiotic resistance landscape. We sequenced and analyzed E. coli adapted to representative antibiotics at increasingly heightened metabolic states. This revealed various underappreciated noncanonical genes, such as those related to central carbon and energy metabolism, which are implicated in antibiotic resistance. These metabolic alterations lead to lower basal respiration, which prevents antibiotic-mediated induction of tricarboxylic acid cycle activity, thus avoiding metabolic toxicity and minimizing drug lethality. Several of the identified metabolism-specific mutations are overrepresented in the genomes of >3500 clinical E. coli pathogens, indicating clinical relevance.
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Mineral oil, BioReagent, Molecular Biology, light oil
AeraSeal film, sterile