Merck
  • Using NMR solvent water relaxation to investigate metalloenzyme-ligand binding interactions.
Using NMR solvent water relaxation to investigate metalloenzyme-ligand binding interactions.
Journal of medicinal chemistry (2009-12-23)
Ivanhoe K H Leung, Emily Flashman, Kar Kheng Yeoh, Christopher J Schofield, Timothy D W Claridge
ABSTRACT
This report demonstrates that solvent water relaxation measurements can be used for quantitative screening of ligand binding and for mechanistic investigations of enzymes containing paramagnetic metal centers by using conventional NMR instrumentation at high field. The method was exemplified using prolyl hydroxylase domain containing enzyme 2 (PHD2), a human enzyme involved in hypoxic sensing, with Mn(II) substituting for Fe(II) at the active site. K(D) values were determined for inhibitors that hinder access of water to the paramagnetic center. This technique is also useful for investigating the mechanism of suitable metalloenzymes, including order of ligand binding and modes of inhibition.
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%