Merck
  • A rapid synthesis of lavendustin-mimetic small molecules by click fragment assembly.
A rapid synthesis of lavendustin-mimetic small molecules by click fragment assembly.
Bioorganic & medicinal chemistry letters (2010-06-24)
Jieun Yoon, Jae-Sang Ryu
ABSTRACT
Lavendustin-mimetic small molecules modifying the linker -CH(2)-NH- with an 1,2,3-triazole ring have been synthesized via a click chemistry. Two pharmacophoric fragments of lavendustin were varied to investigate chemical space and the auxophoric -CH(2)-NH- was altered to an 1,2,3-triazole for rapid click conjugation. The small molecules were evaluated against HCT116 colon cancer and CCRF-CEM leukemia cell lines. Among 28 analogues, 3-phenylpropyl ester 26b inhibited CCRF-CEM leukemia cell growth with GI(50) value of 0.9microM.